Anti-NSRP1 Chicken Polyclonal Antibody
Catalog # ABNOPAB25732
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Speckle Splicing Regulatory Protein 1
- Antigen symbol:NSRP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- ImmunoFluorescence:Yes
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:84081
- Antigen synonyms:NSRP70|Coiled-coil domain-containing protein 55|Nuclear speckle splicing regulatory protein 1|NSrp70|HSPC095|CCDC55
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids at C-terminus of human NSRP1.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Chicken polyclonal antibody raised against synthetic peptide of NSRP1.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: NSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat