Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25032
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Activating transcription factor 4
- Antigen symbol:ATF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:468
- Antigen synonyms:DNA-binding protein TAXREB67|cAMP-dependent transcription factor ATF-4|Cyclic AMP-dependent transcription factor ATF-4|TXREB|TaxREB67|Tax-responsive enhancer element-binding protein 67|CREB2|Activating transcription factor 4|cAMP-responsive element-binding protein 2|CREB-2|Cyclic AMP-responsive element-binding protein 2|ATF4
- Modification:phospho-Ser245
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S245 of ATF4.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ATF4.
- Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200; Immunoprecipitation: 1:50-1:200
Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human