Anti-CYP2C8 Rabbit Polyclonal Antibody
ABNOPAB24906
: Abnova
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily C, Polypeptide 8
- Antigen symbol:CYP2C8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:1558|1559|1562|1557
- Antigen synonyms:P450 form 1|CYPIIC8|S-mephenytoin 4-hydroxylase|CPC8|P450 IIC2|P450 MP-12/MP-20|MP-12/MP-20
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to CYP2C8/CYP2C9/CYP2C18/CYP2C19.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of CYP2C8/CYP2C9/CYP2C18/CYP2C19.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200
Type: Primary
Antigen: CYP2C8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human