Order Entry
Italy
ContactUsLinkComponent
Anti-EPCAM Mouse Monoclonal Antibody
Anti-EPCAM Mouse Monoclonal Antibody
Catalog # ABNOMAB11144
Supplier:  Abnova
Anti-EPCAM Mouse Monoclonal Antibody
Catalog # ABNOMAB11144
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Epithelial cell adhesion molecule
  • Antigen symbol:
    EPCAM
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4072
  • Antigen synonyms:
    CD326|TACSTD1|MIC18|DIAR5|M4S1|EGP40|EGP314|KS1/4|KSA|MK-1|TROP1|HNPCC8|ESA|EGP-2
  • Storage buffer:
    PBS, pH 7,4 (1% BSA, 50% glycerol, 0,02% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to N-terminus of human EPCAM.
  • Purification:
    Protein G purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant protein of EPCAM.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:100-1:2000

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human