Anti-FOSB Mouse Monoclonal Antibody [clone: 83B1138]
ABNOMAB0083
: Abnova
- Antibody type:Primary
- Antigen name:FBJ murine osteosarcoma viral oncogene homolog B
- Antigen symbol:FOSB
- Clonality:Monoclonal
- Clone:83B1138
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2354
- Antigen synonyms:AP-1|GOSB|G0S3|GOS3
- Amino acid number:2 to 15
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 2-15 of human FOSB.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of FOSB.
Type: Primary
Antigen: FOSB
Clonality: Monoclonal
Clone: 83B1138
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human