Anti-ADRA2B Mouse Monoclonal Antibody [clone: 5G10]
ABNOMAB1808
: Abnova
- Antibody type:Primary
- Antigen name:Adrenoceptor Alpha 2B
- Antigen symbol:ADRA2B
- Clonality:Monoclonal
- Clone:5G10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:24174
- Antigen synonyms:ADRA2L1|FAME2|ALPHA2BAR|alpha-2BAR|ADRARL1|ADRA2RL1|ADRA2B
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to third intracellular loop of rat Adra2b.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of Adra2b.
Type: Primary
Antigen: ADRA2B
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat