Anti-CASP14 Rabbit Polyclonal Antibody
ABNOPAB0250
: Abnova
- Antibody type:Primary
- Antigen name:Caspase 14
- Antigen symbol:CASP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Gerbil,Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:23581
- Antigen synonyms:Mini-ICE|Caspase-14|mini-ICE|CASP-14|MICE
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human CASP14.
- Size:50 μl
- Pk:50 µl
Rabbit polyclonal antibody raised against full length recombinant CASP14.
Type: Primary
Antigen: CASP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gerbils, Human, Mouse, Rat