Anti-DAXX Rabbit Polyclonal Antibody
ABNOPAB0254
: Abnova
- Antibody type:Primary
- Antigen name:Death-associated protein 6
- Antigen symbol:DAXX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Monkey,Human,Bovine,Dog,Rat
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:1616
- Antigen synonyms:Fas death domain-associated protein|MGC126246|BING2|EAP1|ETS1-associated protein 1|DAP6|MGC126245|Death domain-associated protein 6|DADB-159G18.9
- Amino acid number:272 to 291
- Storage buffer:0,05% sodium azide
- Sequence:ERLINKPGPDTFPDYGDVLR
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 272-291 of human DAXX.
- Size:50 μl
- Pk:50 µl
Rabbit polyclonal antibody raised against synthetic peptide of DAXX.
Type: Primary
Antigen: DAXX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Monkey, Rat