Anti-SATB2 Rabbit Polyclonal Antibody
ABNOPAB13378
: Abnova
- Antibody type:Primary
- Antigen name:SATB homeobox 2
- Antigen symbol:SATB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:23314
- Antigen synonyms:FLJ21474|MGC119477|MGC119474|SATB family member 2|DNA-binding protein SATB2|FLJ32076|special AT-rich sequence-binding protein 2|KIAA1034
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 14 amino acids of human SATB2.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of SATB2.
Recommended Dilutions: Western Blot: 2-4 ?g/ml
Type: Primary
Antigen: SATB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat