Anti-POU2F1 Rabbit Polyclonal Antibody
ABNOPAB12771
: Abnova
- Antibody type:Primary
- Antigen name:POU Class 2 Homeobox 1
- Antigen symbol:POU2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:5451|18986|171068
- Antigen synonyms:OTF-1|class 2|octamer-binding protein 1|OCT1|transcription factor 1|OTF1|oct-1B|NF-A1|oct-1|POU domain|octamer-binding transcription factor 1|Octamer-binding transcription factor-1
- Storage buffer:PBS, pH 7,2 (10% Proclin300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of human, mouse and rat POU2F1.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of POU2F1.
Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 0.5-2 ?g/ml
Type: Primary
Antigen: POU2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat