Anti-GLB1 Mouse Monoclonal Antibody [clone: DC1 4C7]
ABNOMAB7238
: Abnova
- Antibody type:Primary
- Antigen name:Galactosidase Beta 1
- Antigen symbol:GLB1
- Clonality:Monoclonal
- Clone:DC1 4C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Bacteria
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:PPCA|PPGB|RP3-337O18.1|NGBE|GLB2|Beta-Gal|GSL
- Storage buffer:PBS
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native from bacteria Beta-Galactosidase.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length Beta-Galactosidase.
Type: Primary
Antigen: GLB1
Clonality: Monoclonal
Clone: DC1 4C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria