Anti-EPB41L3 Rabbit Polyclonal Antibody
ABNOPAB15748
: Abnova
- Antibody type:Primary
- Antigen name:Erythrocyte membrane protein band 4.1-like 3
- Antigen symbol:EPB41L3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:116724
- Antigen synonyms:differentially expressed in adenocarcinoma of the lung|EPB41L3|DAL-1|erythrocyte membrane protein band 4.1-like 3 |HGNC:3380|FLJ37633|DAL1|4.1B|KIAA0987
- Storage buffer:10 mM phosphate buffer, 150 mM NaCl, pH 7,4 (0,05% sodium azide)
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with 200 ?l distilled water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to 197 amino acids (P790-L986) of rat Epb4.1l3.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against partial recombinant Epb4.1l3.
Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:25
Type: Primary
Antigen: EPB41L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat