Anti-ARIH1 Goat Polyclonal Antibody
Catalog # ABNOPAB6322
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ariadne RBR E3 ubiquitin protein ligase 1
- Antigen symbol:ARIH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:25820
- Antigen synonyms:ARIH1, HHARI, ARI, HARI, UBCH7BP, ariadne homolog, ubiquitin-conjugating enzyme E2 binding protein, 1 (Drosophila), ariadne homolog, ariadne, Drosophila, homolog of, DKFZp686O13120, FLJ20329, FLJ93118, ariadne ubiquitin-conjugating enzyme E2 binding protein homolog 1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-HEGYEKDLWEYIED
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human ARIH1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of ARIH1.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.2-1 ?g/ml
Type: Primary
Antigen: ARIH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human