Anti-CTR9 Rabbit Polyclonal Antibody
Catalog # ABNOPAB14078
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CTR9 homolog, Paf1/RNA polymerase II complex component
- Antigen symbol:CTR9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:22083
- Antigen synonyms:p150TSP|p150|SH2BP1|TSBP
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus of mouse Ctr9.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Ctr9.
Recommended Dilutions: ELISA: 1:100-1:500; Western Blot: 1:500
Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse