Anti-GRB10 Rabbit Polyclonal Antibody
ABNOPAB18386
: Abnova
- Antibody type:Primary
- Antigen name:Growth factor receptor-bound protein 10
- Antigen symbol:GRB10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2887
- Antigen synonyms:KIAA0207|GRBIR|GRB10 adapter protein|Insulin receptor-binding protein Grb-IR|GRB10
- Storage buffer:PBS, 150mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues surrounding Y67 of human GRB10.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of GRB10.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:5000
Type: Primary
Antigen: GRB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human