Anti-PCNX Goat Polyclonal Antibody
ABNOPAB6003
: Abnova
- Antibody type:Primary
- Antigen name:pecanex homolog (Drosophila)
- Antigen symbol:PCNX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:22990
- Antigen synonyms:PCNXL1|pecanex
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-TVIETGVLELGAEV
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human PCNX.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of PCNX.
Recommended Dilutions: ELISA: 1:64000
Type: Primary
Antigen: PCNX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: