Anti-GPI Goat Polyclonal Antibody
ABNOPAB17845
: Abnova
- Antibody type:Primary
- Antigen name:Glucose-6-phosphate isomerase
- Antigen symbol:GPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:2821
- Antigen synonyms:glucose phosphate isomerase|oxoisomerase|PGI|GNPI|hexosephosphate isomerase|AMF|autocrine motility factor|PHI|phosphoglucose isomerase|sperm antigen-36|NLK|glucose-6-phosphate isomerase|phosphohexomutase|phosphohexose isomerase|phosphosaccharomutase|neuroleukin|GPI|SA-36|hexose monophosphate isomerase
- Storage buffer:0,5 mg/mL Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
- Sequence:C-YREHRSELNLRR
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at internal region of human GPI.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of GPI.
Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.01-0.03 ?g/ml
Type: Primary
Antigen: GPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human