Order Entry
Italy
ContactUsLinkComponent
Anti-OPRM1 Rabbit Polyclonal Antibody
Anti-OPRM1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18103
Supplier:  Abnova
Anti-OPRM1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18103
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Opioid Receptor, Mu 1
  • Antigen symbol:
    OPRM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4988
  • Antigen synonyms:
    mor|MOR-1O|M-OR-1|MOP-R|muOR|MOR-1|Oprm
  • Storage buffer:
    PBS, 150mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human OPRM1.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of OPRM1.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100; ELISA: 1:20000

Type: Primary
Antigen: OPRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat