Anti-ADGRL2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25951
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adhesion G Protein-Coupled Receptor L2
- Antigen symbol:ADGRL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Gorilla,Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:23266
- Antigen synonyms:LPHH1|LPHN2|LEC1|CL2|CIRL2
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids at C-terminus of human LPHN2.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of LPHN2.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 14 ?g/ml
Type: Primary
Antigen: ADGRL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human