Anti-FAM63B Rabbit Polyclonal Antibody
ABGEAP13388B
: Abgent
- Antibody type:Primary
- Antigen name:Family with sequence similarity 63, member B
- Antigen symbol:FAM63B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Gene ID:54629
- Amino acid number:577 to 606
- Storage buffer:pH 7,4
- Molecular weight:67.106 kDa
- Sequence:RQSGNSERKRKEPREC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.81
- Shipping temperature:4 °C
- Immunogen:This FAM63B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 577-606 amino acids from the C-terminal region of human FAM63B.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: FAM63B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human