Order Entry
Italy
ContactUsLinkComponent
 
Anti-ARHGEF6 Rabbit Polyclonal Antibody
  ABGEAP16738B
 :  Abgent
Anti-ARHGEF6 Rabbit Polyclonal Antibody
  ABGEAP16738B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Rac/Cdc42 guanine nucleotide exchange factor (GEF) 6
  • Antigen symbol:
    ARHGEF6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Gene ID:
    9459
  • Antigen synonyms:
    MRX46|PIXA|alpha-PIX|COOL2|alphaPIX|Cool-2
  • Amino acid number:
    614 to 641
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    87.499 kDa
  • Sequence:
    FLHKRKTERKPSEC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.120
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ARHGEF6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 614-641 amino acids from the C-terminal region of human ARHGEF6.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: ARHGEF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human