Order Entry
Italy
ContactUsLinkComponent
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: P4D1]
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: P4D1]
Supplier:  US Biological
Certificates
undefined
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: P4D1]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIU1000-07J-25
  • USBIU1000-07J-100
  • Antigen symbol:
  • Ubi-1
  • Ubi-1
  • Antigen name:
  • Ubiquitin
  • Ubiquitin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • P4D1
  • P4D1
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Immunogen:
  • Denatured glutaraldehyde cross-linked ubiquitin IgG complex
  • Denatured glutaraldehyde cross-linked ubiquitin IgG complex
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by immunoaffinity chromatography
  • purified by immunoaffinity chromatography
  • Concentration:
  • ~0,5 mg/ml
  • ~0,5 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,2, 0,09% sodium azide.
  • PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together