Anti-GPC1 Rabbit Polyclonal Antibody
ORIGTA319424
: OriGene
- Antibody type:Primary
- Antigen symbol:GPC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Cross adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Anti-Glypican-1 protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human glypican-1 protein.
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: GPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat