Anti-TREM2 Rat Monoclonal Antibody [clone: 13A84]
USBIT8282-11D
- Antibody type:Primary
- Antigen name:Triggering receptor expressed on myeloid cells 2
- Antigen symbol:TREM2
- Clonality:Monoclonal
- Clone:13A84
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Recommended dilution: Flow Cytometry: 1:50
Type: Primary
Antigen: TREM2 (triggering receptor expressed on myeloid cells 2)
Clonality: monoclonal
Clone: 13A84
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse