Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Anti-MPO Rabbit Polyclonal Antibody

Anti-MPO Rabbit Polyclonal Antibody

100 µg
Catalog # USBIM9760-06F
Supplier:   US Biological
Anti-MPO Rabbit Polyclonal Antibody
Catalog # USBIM9760-06F
Supplier:  US Biological
Anti-MPO Rabbit Polyclonal Antibody

100 µg

Price
463,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Specifications
  • Taglia:
    100 µg
  • Western blot:
    Yes
  • Reattività:
    Human|Rat|Mouse
  • Purificazione:
    purified by affinity chromatography
  • Tampone di conservazione:
    PBS, 0,5% BSA, 5mM EDTA, 0,01% thimerosal,30% glycerol.
  • Coniugazione:
    Non coniugato
  • Isotipo:
    IgG
  • Cat. No.:
    USBIM9760-06F
  • Forma:
    liquid
  • Simbolo dell'antigene:
    MPO
  • Clonalità:
    Policlonale
  • Ospite:
    Rabbit
  • Temperatura di spedizione:
    Ice
  • Tipo anticorpo:
    Primary
  • Temperatura di conservazione:
    Cool
  • Tipo di anticorpi:
    Myeloperoxidase
  • Concentrazione:
    ~0,5 mg/ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.