Anti-Macrophage Scavenger Receptor Type I Rat Monoclonal Antibody [clone: 10B1015]
Catalog # USBIM1210-04A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Macrophage Scavenger Receptor Type I
- Clonality:Monoclonal
- Clone:10B1015
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Pig,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50-1:100
Type: Primary
Antigen: Macrophage Scavenger Receptor Type I
Clonality: monoclonal
Clone: 10B1015
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Pig