Anti-KDM4A Rabbit Polyclonal Antibody
Catalog # USBIJ7876-45G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:lysine (K)-specific demethylase 4A
- Antigen symbol:KDM4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: KDM4A (lysine (K)-specific demethylase 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human