Anti-HMGA2 Rabbit Polyclonal Antibody
Catalog # USBIH3869-51A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:High mobility group AT-hook 2
- Antigen symbol:HMGA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Storage buffer:10mM PBS, pH 7,4, BSA, sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 15 ?g/ml
Type: Primary
Antigen: HMGA2 (high mobility group AT-hook 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human