Anti-GABA Receptor Associated Protein Rabbit polyclonal antibody
Catalog # USBIG1015-24E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:GABA Receptor Associated Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:500-1:1000
Type: Primary
Antigen: GABA Receptor Associated Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat