Order Entry
Italy
ContactUsLinkComponent
 
Anti-GABPB1 Goat polyclonal antibody
Catalog # USBIG1009-84A
Supplier:  US Biological
undefined
Anti-GABPB1 Goat polyclonal antibody
Catalog # USBIG1009-84A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GA Binding Protein Transcription Factor, Beta Subunit 1
  • Antigen symbol:
    GABPB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    internal
  • Form:
    liquid
  • Storage buffer:
    Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-1. 5 ?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: GABPB1 (GA binding protein transcription factor, beta subunit 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human