Anti-Factor V, activation peptide, 120kD Mouse Monoclonal Antibody [clone: 3H367]
Catalog # USBIF0011-23
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Factor V, activation peptide, 120kD
- Clonality:Monoclonal
- Clone:3H367
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Molecular weight:120 kDa
- Storage temperature:Cool
- Concentration:~4 mg/ml
- Shipping temperature:Ice
- Tested applications:Radio Immunoassay
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 5 ?g/ml
Type: Primary
Antigen: Factor V, activation peptide, 120kD
Clonality: monoclonal
Clone: 3H367
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human