Anti-THY1 Mouse Monoclonal Antibody [clone: F15-42-1]
USBIC2441-02F
- Antibody type:Primary
- Antigen name:Thy1 Cell Surface Antigen
- Antigen symbol:THY1
- Clonality:Monoclonal
- Clone:F15-42-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:200 µg
- Pk:200 µG
Recommended dilution: Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: Monoclonal
Clone: F15-42-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human