Anti-FCGR2B Goat polyclonal antibody
USBIC2384-03B
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, low affinity 2B, receptor (CD32)
- Antigen symbol:FCGR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 0. 3-1 ?g/ml; ELISA: 1:16000; Immunochemistry: 3. 75 ?g/ml
Type: Primary
Antigen: FCGR2B (Fc fragment of IgG, low affinity IIb, receptor (CD32))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human