Anti-CDH17 Mouse Monoclonal Antibody [clone: 1H3]
USBIC0107-17
- Antibody type:Primary
- Antigen name:cadherin 17, LI cadherin (liver-intestine)
- Antigen symbol:CDH17
- Clonality:Monoclonal
- Clone:1H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:24 to 132
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: CDH17 (cadherin 17, LI cadherin (liver-intestine))
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human