Anti-ADAMTS13 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
USBIA0859-61Z1-AP
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase with thrombospondin type 1 motif, 13
- Antigen symbol:ADAMTS13
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Form:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: ADAMTS13 (ADAM metallopeptidase with thrombospondin type 1 motif, 13)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human