Anti-IL25 Goat Polyclonal Antibody
Catalog # USBI167429
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 25
- Antigen symbol:IL25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
- Sequence:RPNRHPESCRASED
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 3-1 ?g/ml
Type: Primary
Antigen: IL25 (interleukin 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human