Anti-CYP26B1 Goat Polyclonal Antibody
USBI150290
- Antibody type:Primary
- Antigen name:cytochrome P450, family 26, subfamily B, polypeptide 1
- Antigen symbol:CYP26B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:128000; Immunochemistry: 4-6?g/ml
Type: Primary
Antigen: CYP26B1 (cytochrome P450, family 26, subfamily B, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse