Order Entry
Italy
ContactUsLinkComponent
 
Anti-HDAC2 Mouse Monoclonal Antibody [clone: 806116]
Catalog # USBI145487
Supplier:  US Biological
undefined
Anti-HDAC2 Mouse Monoclonal Antibody [clone: 806116]
Catalog # USBI145487
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Histone deacetylase 2
  • Antigen symbol:
    HDAC2
  • Clonality:
    Monoclonal
  • Clone:
    806116
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS,Trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A/G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.2?g/ml

Type: Primary
Antigen: HDAC2 (histone deacetylase 2)
Clonality: monoclonal
Clone: 806116
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat