Anti-Caspr1 Sheep Polyclonal Antibody
Catalog # USBI145406
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Caspr1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS,Trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
- Recommended dilution: Immunochemistry: 515 μg/ml
Type: Primary
Antigen: Caspr1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human