Order Entry
Italy
ContactUsLinkComponent
Anti-CHD4 Mouse Monoclonal Antibody [clone: 13k32]
Anti-CHD4 Mouse Monoclonal Antibody [clone: 13k32]
Catalog # USBI143159
Supplier:  US Biological
undefined
Anti-CHD4 Mouse Monoclonal Antibody [clone: 13k32]
Catalog # USBI143159
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromodomain helicase DNA binding protein 4
  • Antigen symbol:
    CHD4
  • Clonality:
    Monoclonal
  • Clone:
    13k32
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,2% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: CHD4 (chromodomain helicase DNA binding protein 4)
Clonality: monoclonal
Clone: 13k32
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human