Order Entry
Italy
ContactUsLinkComponent
Anti-SYNM Mouse Monoclonal Antibody [clone: 4G5]
Anti-SYNM Mouse Monoclonal Antibody [clone: 4G5]
  USBI134141
Anti-SYNM Mouse Monoclonal Antibody [clone: 4G5]
  USBI134141

 

  • Antibody type:
    Primary
  • Antigen name:
    synemin, intermediate filament Protein
  • Antigen symbol:
    SYNM
  • Clonality:
    Monoclonal
  • Clone:
    4G5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Format:
    ascites
  • Form:
    liquid
  • Amino acid number:
    1466 to 1566
  • Storage buffer:
    ascites fluid.
  • Sequence:
    VSNVEAIRSRTQEAGALGVSDRGSWRDADSRNDQAVGVSFKASAGEGDQAHREQGKEQAMFDKKVQLQRMVDQRSVISDEKKVALLYLDNEEEENDGHWF*
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: SYNM (synemin, intermediate filament protein)
Clonality: monoclonal
Clone: 4G5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human