Anti-STAU1 Mouse Monoclonal Antibody [clone: 3D8]
USBI133946
- Antibody type:Primary
- Antigen name:Staufen double-stranded RNA binding protein 1
- Antigen symbol:STAU1
- Clonality:Monoclonal
- Clone:3D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:401 to 497
- Storage buffer:PBS, pH 7,2.
- Sequence:PHGPLTRPSEQLDYLSRVQGFQVEYKDFPKNNKNEFVSLINCSSQPPLISHGIGKDVESCHDMAALNILKLLSELDQQSTEMPRTGNGPMSVCGRC
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: STAU1 (staufen double-stranded RNA binding protein 1)
Clonality: monoclonal
Clone: 3D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human