Anti-LONP1 Mouse Monoclonal Antibody [clone: 3B2]
USBI129176
- Antibody type:Primary
- Antigen name:lon peptidase 1, mitochondrial
- Antigen symbol:LONP1
- Clonality:Monoclonal
- Clone:3B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:661 to 761
- Storage buffer:PBS, pH 7,2.
- Sequence:YVAQEKLAIAERYLVPQARALCGLDESKAKLSSDVLTLLIKQYCRESGVRNLQKQVEKVLRKSAYKIVSGEAESVEVTPENLQDFVGKPVFTVERMYDVTP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: LONP1 (lon peptidase 1, mitochondrial)
Clonality: monoclonal
Clone: 3B2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human