Order Entry
Italy
ContactUsLinkComponent
Anti-ATG16L1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 54CT27.2.6]
Anti-ATG16L1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 54CT27.2.6]
Catalog # USBI030790-FITC
Supplier:  US Biological
Anti-ATG16L1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 54CT27.2.6]
Catalog # USBI030790-FITC
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    autophagy related 16-like 1
  • Antigen symbol:
    ATG16L1
  • Clonality:
    Monoclonal
  • Clone:
    54CT27.2.6
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:4000; Western Blot: 1:60; Immunochemistry: 1:10-50

Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality: Monoclonal
Clone: 54CT27.2.6
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human