Anti-ERBB2 Mouse Monoclonal Antibody [clone: 13B373]
Catalog # USBI030295
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2
- Antigen symbol:ERBB2
- Clonality:Monoclonal
- Clone:13B373
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ERBB2 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2)
Clonality: Monoclonal
Clone: 13B373
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human