Anti-GOX Rabbit Polyclonal Antibody
Catalog # ROCK100-4137
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glucose Oxidase
- Antigen symbol:GOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Aspergillus niger
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
- Storage temperature:Cool
- Concentration:90 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Glucose Oxidase (Aspergillus niger)
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Specifications
About this item
Anti-Glucose Oxidase Antibody has been assayed against 1.0 µg of Glucose Oxidase (Aspergillus niger) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000
Restore with deionized water (or equivalent); 2.0 ml
Type: Primary
Antigen: GOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Aspergillus niger