Order Entry
Italy
ContactUsLinkComponent
Anti-Glial Fibrillary Acidic Protein (GFAP) (G-A-5) Mouse Monoclonal Antibody
Anti-Glial Fibrillary Acidic Protein (GFAP) (G-A-5) Mouse Monoclonal Antibody
  IF03L-100
Anti-Glial Fibrillary Acidic Protein (GFAP) (G-A-5) Mouse Monoclonal Antibody
  IF03L-100

 

  • Antibody type:
    Primary
  • Antigen name:
    Glial fibrillary acidic protein
  • Antigen symbol:
    GFAP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: Glial Fibrillary Acidic Protein (GFAP) (G-A-5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: