Order Entry
Italy
ContactUsLinkComponent
 
Anti-FGF14 Goat Polyclonal Antibody
  EVBIEB11834
undefined
Anti-FGF14 Goat Polyclonal Antibody
  EVBIEB11834

 

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor 14
  • Antigen symbol:
    FGF14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: FGF14 (fibroblast growth factor 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: