Anti-Caveolin-2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1540
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Caveolin 2
- Antigen symbol:CAV2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CAV2|CAV|caveolin 2 isoform a and b|caveolin-2|20-kD|caveolae protein|Caveolin-2
- Amino acid number:1 to 17
- Sequence:MGLETEKADVQLFMDDD
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Caveolin-2 (1-17aa MGLETEKADVQLFMDDD), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Caveolin-2(CAV2) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Caveolin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat