Anti-ZBTB7A Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-23673
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 7A
- Antigen symbol:ZBTB7A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:51341
- Antigen synonyms:pokemon|ZNF857A|FBI-1|LRF|ZBTB7|FBI1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: ZBTB7A (zinc finger and BTB domain containing 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: